Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P3018

Sigma-Aldrich

L-Phenylalanine-Agarose

saline suspension

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
23151817
NACRES:
NA.56

form

saline suspension

extent of labeling

2-10 μmol per mL

matrix

Cross-linked 4% beaded agarose

matrix activation

cyanogen bromide

matrix attachment

amino

matrix spacer

1 atom

storage temp.

2-8°C

Application

L-Phenylalanine-agarose is used in protein chromatography, affinity chromatography and amino acid resins. L-Phenylalanine-agarose has been used to purify and characterize a cysteine protease of Fasciola gigantica adult worms as well as to purify a proteolytic enzyme of the liver fluke Fasciola species.

Physical form

Suspension in 2.0 M NaCl containing preservative

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Aoki et al.
Molecular and biochemical parasitology, 8(2), 89-97 (1983-06-01)
A proteolytic enzyme of the liver fluke Fasciola sp. was purified as described previously by ammonium sulfate fractionation, gel filtration on Sephadex G-200 column and L-phenylalanine-agarose chromatography. Leupeptin, a peptide aldehyde of microbial origin, competitively inhibited the enzyme activity with
J F Geissler et al.
Journal of bacteriology, 170(4), 1709-1714 (1988-04-01)
A soluble benzoate-coenzyme A (CoA) ligase was purified from the phototrophic bacterium Rhodopseudomonas palustris. Synthesis of the enzyme was induced when cells were grown anaerobically in light with benzoate as the sole carbon source. Purification by chromatography successively on hydroxylapatite
K H Lau et al.
The Journal of biological chemistry, 260(8), 4653-4660 (1985-04-25)
A partially purified bovine cortical bone acid phosphatase, which shared similar characteristics with a class of acid phosphatase known as tartrate-resistant acid phosphatase, was found to dephosphorylate phosphotyrosine and phosphotyrosyl proteins, with little activity toward other phosphoamino acids or phosphoseryl
B O Fagbemi et al.
Veterinary parasitology, 43(3-4), 223-232 (1992-07-01)
A 26-28 kDa protease was isolated from Fasciola gigantica adult worms by a two-stage purification process of column chromatography in a Sephacryl S-200 column and affinity chromatography in an L-phenylalanine-agarose column. This protease is a cysteine (thiol) proteinase with an
H R Onishi et al.
The Journal of biological chemistry, 254(23), 11943-11952 (1979-12-10)
The nonspecific alkaline phosphatase of yeast (Saccharomyces strain 1710) has been purified by ion exchange, hydrophobic, and affinity chromatography. This vacuolar enzyme has a molecular weight of 130,000 and is composed of subunits (probably of 66,000 molecular weight). It also

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service