Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P2905

Sigma-Aldrich

[Nle8,18, Tyr34]-PTH Fragment 1-34 amide bovine

≥97% (HPLC)

Synonym(s):

Ala-Val-Ser-Glu-Ile-Gln-Phe-Nle-His-Asn-Leu-Gly-Lys-His-Leu-Ser-Ser-Nle-Glu-Arg-Val-Glu-Trp-Leu-Arg-Lys-Lys-Leu-Gln-Asp-Val-His-Asn-Tyr-NH2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C185H293N55O50
CAS Number:
Molecular Weight:
4087.64
MDL number:
UNSPSC Code:
12352200

assay

≥97% (HPLC)

form

powder

storage temp.

−20°C

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Lafond et al.
Endocrinology, 123(6), 2834-2840 (1988-12-01)
The syncytiotrophoblast of the placenta is the site of exchange of nutrients and minerals between the mother and fetus. We have recently demonstrated that PTH influences, in vitro, phosphate transport through the placenta brush border membranes (BBM) and increases cAMP
I Yamamoto et al.
Endocrinology, 122(4), 1208-1217 (1988-04-01)
PTH receptors on two stable clonal rat osteosarcoma cell lines, ROS 17/2 and ROS 17/2.8, were characterized using an HPLC-purified, synthetic, sulfur-free, radioiodinated analog of bovine PTH, [Nle8,Nle18,Tyr,34]bovine PTH-(1-34)amide. PTH binding is specific for PTH agonists and antagonists and is
L G Rao et al.
Endocrinology, 117(4), 1632-1638 (1985-10-01)
Most studies of PTH receptor binding have been carried out with amino-terminal radioligands which only detect binding within that region of the hormone molecule. We studied the binding of electrolytically labeled intact bovine PTH [bPTH-(1-84)], and its amino-terminal fragment, bPTH-(1-34)
H M Perry et al.
The Journal of biological chemistry, 259(9), 5531-5535 (1984-05-10)
In this article we demonstrate receptors for parathyroid hormone in circulating mononuclear leukocytes using the radioiodinated analogue (8,18 norleucine, 34 tyrosine) bPTH 1-34 (bovine parathyroid hormone 1-34). Specific binding, which is reversible and saturable, equilibrates within 5 min at 0-4
P D'Amour et al.
The American journal of physiology, 246(3 Pt 1), E249-E255 (1984-03-01)
The fractional hepatic extraction (FHE) of oxidized 125iodine-labeled bovine parathyroid hormone, 125I-bPTH-(1-84), is dependent on the integrity of the 28-48 sequence of the bPTH structure. A second type of FHE, related to the biologically active core of the hormone, is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service