Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P2803

Sigma-Aldrich

N-(6-Phenylhexyl)-5-chloro-1-naphthalenesulfonamide

Synonym(s):

SC-9

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H24ClNO2S
CAS Number:
Molecular Weight:
401.95
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

form

solid

storage temp.

−20°C

SMILES string

Clc1cccc2c(cccc12)S(=O)(=O)NCCCCCCc3ccccc3

InChI

1S/C22H24ClNO2S/c23-21-15-8-14-20-19(21)13-9-16-22(20)27(25,26)24-17-7-2-1-4-10-18-11-5-3-6-12-18/h3,5-6,8-9,11-16,24H,1-2,4,7,10,17H2

InChI key

YUXMTDYHFPRPLG-UHFFFAOYSA-N

Biochem/physiol Actions

Activates protein kinase C in a Ca2+-dependent manner

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Hirobe
Journal of cell science, 107 ( Pt 6), 1679-1686 (1994-06-01)
Mouse epidermal melanoblasts preferentially proliferated from disaggregated epidermal cell suspensions derived from newborn mouse skin in a serum-free melanoblast proliferation medium containing dibutyryl adenosine 3':5'-cyclic monophosphate and basic fibroblast growth factor. After 12 days, almost all of the keratinocytes died
B Petersen et al.
Cell biology and toxicology, 10(4), 219-230 (1994-08-01)
TGF-beta 1 is known as a potent inhibitor of proliferation of rat and human hepatocytes. In this study we show that the effects of TGF-beta 1 are quite different on mouse hepatocytes. In rat and human hepatocytes, TGF-beta 1 inhibited
K Ashizawa et al.
Reproduction (Cambridge, England), 131(6), 1017-1024 (2006-06-01)
The signal transduction pathways involved in the regulation of the acrosome reaction and motility of fowl spermatozoa were investigated. The motility and acrosomal integrity of fowl spermatozoa in TES/NaCl buffer, with or without homogenised inner perivitelline layers (IPVL), prepared from
Mohamed I Husseiny et al.
Vaccine, 27(28), 3780-3787 (2009-05-26)
Salmonella enterica is a versatile live carrier for the presentation of recombinant vaccine antigens. Fusion proteins of a type III secretion system effector and heterologous vaccine antigens can be translocated by live attenuated Salmonella strains and mediate protective immunity against
M Kavaliers et al.
Brain research, 554(1-2), 65-71 (1991-07-19)
There is substantial evidence that magnetic fields can reduce opiate-induced analgesia, with alterations in calcium channel function and/or calcium ion flux being implicated in the mediation of these inhibitory effects. The present experiments were designed to examine the effects of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service