Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P2743

Sigma-Aldrich

Pyruvate Dehydrogenase Phosphatase from bovine kidney

buffered aqueous glycerol solution, ~850 units/mg protein

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.32

form

buffered aqueous glycerol solution

Quality Level

specific activity

~850 units/mg protein

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

General description

Pyruvate dehydrogenase phosphatase (PDP) (EC 3.1.3.43) contains two genetically and biochemically different isoforms, PDP1 and PDP2.[1] PDP1 is widely distributed and is composed of two subunits PDP1c, a catalytic subunit and PDP1r, a flavoprotein with a bound FAD.[2][3]

Biochem/physiol Actions

Insulin-activated Mg2+-dependent and Ca2+-stimulated mitochondrial protein phosphatase specific for the pyruvate dehydrogenase complex.
Pyruvate dehydrogenase phosphatase catalytic subunit 1 (PDP1) catalyzes a Mg2+,-Ca2+ mediated dephosphorylation and activation of E1.[2] PDP1 plays a crucial role in the regulation of pyruvate dehydrogenase complex (PDC) activity.[1]

Unit Definition

One unit will releases 1 nmol of inorganic phosphate from 32P-labeled pyruvate dehydrogenase complex per min at 30 °C, pH 7.0.

Physical form

Solution in 50 μl of 50 mM Tris-HCl, pH 7.0, containing 14 mM β-mercaptoethanol, 1 mM benzamidine, 0.1 mM PMSF, 1 mM EDTA, and 50% glycerol.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Sens. 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structural Requirements within the Lipoyl Domain for the Ca2+-dependent Binding and Activation of Pyruvate Dehydrogenase Phosphatase Isoform 1 or Its Catalytic Subunit
Turkan A, et al.
The Journal of Biological Chemistry, 277, 14976-14985 (2002)
Purification of Bovine Kidney and Heart Pyruvate Dehydrogenaseb Phosphatase on Sepharose Derivatized with the Pyruvate Dehydrogenase Complex
Pratt ML, et al.
European Journal of Biochemistry, 125, 349-355 (1982)
Down?regulation of Pyruvate Dehydrogenase Phosphatase in Obese Subjects is a Defect that Signals Insulin Resistance
Piccinini M, et al.
Obesity Research, 13, 678-686 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service