Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P2737

Greiner PCR 96 well plates, polycarbonate

Type 5 for Techne, 96 well

Synonym(s):

96 multiwell plates, 96 well microplates, 96 well microtiter plates, 96 well plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300

material

polycarbonate, type 5

sterility

non-sterile

packaging

case of 50

manufacturer/tradename

Greiner 651580

technique(s)

PCR: suitable

color

transparent plate

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

PCR Plates have a standard microplate footprint and are compatible with thermocyclers of most major brands. DNAse and RNAse free and non-pyrogenic by LAL method to <0.06 EU/ml.  
ThermoQuick 96W PCR Microplate, well working volume 100 μL.

Other Notes

96 well microplate, without lid, solid round (U) bottom well design, alphanumeric well coding, length: 127.76+/- 0.15 mm width: 85.48 +/- 0.15 mm curvature: approx.200 μm.

Suitability

Use for polymerase chain reaction; suitable Techne-96/ Eppendorf Gradient, optimal and rapid heat transmission; Free of detectable DNase/RNase, human DNA and pyrogens.

Legal Information


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service