P2612
Greiner PCR 96 well plates, polycarbonate
Type 4 for PerkinElmer and Thermolyne, 96 well
Synonym(s):
96 multiwell plates, 96 well microplates, 96 well microtiter plates, 96 well plates
About This Item
Recommended Products
material
polycarbonate, type 4
sterility
non-sterile
packaging
case of 50
manufacturer/tradename
Greiner 651570
technique(s)
PCR: suitable
color
transparent plate
binding type
non-treated surface
Looking for similar products? Visit Product Comparison Guide
General description
Other Notes
Suitability
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service