Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P2490

Sigma-Aldrich

Presenilin-1 N-Terminal Peptide

lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202

form

lyophilized powder

species reactivity

human

concentration

≥50% (HPLC)

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... PSEN1(5663)

Amino Acid Sequence

Gln-Met-Ser-Glu-Asp-Asn-His-Leu-Ser-Asn-Thr-Val-Arg-Ser-Gln-Asn-Asp-Asn-Arg

Biochem/physiol Actions

Peptide used to study effects on the production of Aβ[X-42] peptide and accumulation of endogenous presenilin.
Used to study production of Aβ[X-42] peptide and accumulation of endogenous presenilin.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service