Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P2487

Greiner PCR 96 well plates, polycarbonate

Type 3 for MJ, Coy, and Stratagene, 96 well

Synonym(s):

96 multiwell plates, 96 well microplates, 96 well microtiter plates, 96 well plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000

material

polycarbonate, type 3

sterility

non-sterile

packaging

case of 50

manufacturer/tradename

Greiner 651560

technique(s)

PCR: suitable

color

transparent plate

Looking for similar products? Visit Product Comparison Guide

General description

PCR Plates have a standard microplate footprint and are compatible with thermocyclers of most major brands. DNAse and RNAse free and non-pyrogenic by LAL method to <0.06 EU/ml.  
ThermoQuick 96W PCR Microplate, well working volume 150 μL.

Other Notes

96 well type 3 microplate, without lid, no skirt alphanumeric well coding, length: 126.50 mm; width: 86; mm thickness 635 μm.

Suitability

Use for polymerase chain reaction; suitable for MJ research, optimal and rapid heat transmission; Free of detectable DNase/RNase, human DNA and pyrogens.

Legal Information


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service