Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P2367

Sigma-Aldrich

PARC−Biotin Conjugate

lyophilized powder

Synonym(s):

AMAC-1, DC-CK1, MIP-4, Pulmonary and Activation-Related

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800

biological source

human

form

lyophilized powder

potency

0.5-2 μg/mL ED50

mol wt

11 kDa

packaging

pkg of 20 μg

storage condition

avoid repeated freeze/thaw cycles

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... CCL18(6362)

Biochem/physiol Actions

Chemoattracts lymphocytes and macrophages, but not monocytes or granulocytes.

Analysis Note

The biological activity is measured by its ability to induce chemotaxis in T-lymphocytes.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Hieshima et al.
Journal of immunology (Baltimore, Md. : 1950), 159(3), 1140-1149 (1997-08-01)
By searching the expressed sequence tag (EST) database, we identified partial cDNA sequences encoding a polypeptide with significant sequence identity to the human CC chemokine macrophage-inflammatory protein-1 alpha (MIP-1 alpha)/LD78 alpha. We determined the complete cDNA sequence that contained a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service