Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P2118

Sigma-Aldrich

Protein Phosphatase 2A Inhibitor Protein, 1‑α human

buffered aqueous glycerol solution

Synonym(s):

I1αPP2A

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352204
NACRES:
NA.32

Product Name

Protein Phosphatase 2A Inhibitor Protein, 1‑α human, recombinant, expressed in E. coli, buffered aqueous glycerol solution

recombinant

expressed in E. coli

Quality Level

form

buffered aqueous glycerol solution

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... SET(6418)

Physical form

Each vial contains 250ng of protein. Supplied as a solution in 50 mM Tris-HCl, pH 7.0 buffer containing 14 mM β-mercaptoethanol, 1 mM benzamidine, 0.1 mM PMSF, 1 mM EDTA, 0.1% Brij® L23 and 10% glycerol.

Legal Information

Brij is a registered trademark of Croda International PLC

pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1 - STOT RE 2 Oral

target_organs

Kidney

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service