Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P1635

Sigma-Aldrich

Petroselinic acid sodium salt

~98%

Synonym(s):

cis-6-Octadecenoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C18H33O2Na
CAS Number:
Molecular Weight:
304.44
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

assay

~98%

form

solid

functional group

carboxylic acid

shipped in

ambient

storage temp.

−20°C

SMILES string

[Na].CCCCCCCCCCC\C=C/CCCCC(O)=O

InChI

1S/C18H34O2.Na.H/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20;;/h12-13H,2-11,14-17H2,1H3,(H,19,20);;/b13-12-;;

InChI key

DFTWBAYJNMSBEU-RWYDNECBSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C E Høy et al.
Lipids, 16(2), 102-108 (1981-02-01)
The incorporation of the dietary cis 18:1(n-12) and cis 18:1(n-10) into liver mitochondrial membrane phospholipids and adipose tissue triglycerides was studied in 4 groups of rats fed diets containing 10 weight percent (wt %) of fat with the following contents
Frederic Destaillats et al.
Lipids, 37(1), 111-116 (2002-03-06)
Dibutyrate derivatives of monoacylglycerols of oleic, petroselinic, and cis-vaccenic acids were prepared by diesterification of monoacylglycerols with n-butyryl chloride. The resulting triacylglycerols were analyzed by gas chromatography (GC) with a 65% phenyl methyl silicone capillary column and separated on the
Kenichi Sakai et al.
Journal of oleo science, 59(10), 541-548 (2010-09-30)
Novel anionic heterogemini surfactants have been synthesized from two kinds of unsaturated fatty acids (oleic acid and petroselinic acid). The hydrocarbon chain is covalently bound to the terminal carbonyl group of the unsaturated fatty acids and hydrophilic headgroups (i.e., sulfonic
N Weber et al.
The Journal of nutrition, 125(6), 1563-1568 (1995-06-01)
Studies in vitro have revealed that triacylglycerols containing petroselinoyl [18:1(n-12)] moieties are hydrolyzed by pancreatic lipase at much lower rates than other triacylglycerols. To assess the lipolysis and absorption in vivo of such unusual triacylglycerols, diets containing 120 g seed
N Tsevegsuren et al.
Lipids, 39(6), 571-576 (2004-11-24)
The occurrence of petroselinic acid (18:1delta6cis) in seed oils was believed to be limited to the Umbelliferae or Apiaceae, and a few other members of the Umbelliflorae. A major occurrence of petroselinic acid outside the Umbelliflorae must therefore be regarded

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service