P1609
Protein Kinase C Catalytic Subunit from rat brain
≥90% (SDS-PAGE), ≥800 units/mg protein
Synonym(s):
PKC-M, PKM
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
assay
≥90% (SDS-PAGE)
form
liquid
specific activity
≥800 units/mg protein
packaging
vial of 200 ng
concentration
25 μg/mL protein
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
rat ... Prkd1(85421)
Biochem/physiol Actions
Does not require Ca2+ or phosphatidylserine for its activity.
Unit Definition
One unit will transfer of 1 nanomole of phosphate per minute at 30 °C using Histone III as substrate and phosphatidylserine as an activator.
Physical form
Storage buffer: 20 mM Tris-HCl, pH 7.5, 2 mM EGTA, 2 mM EDTA, 1 mM DTT, 10 mM potassium phosphate, 50% glycerol, 0.05% Triton™ X-100
Preparation Note
Prepared by tryptic digestion of PKC.
Legal Information
Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow
Storage Class
10 - Combustible liquids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
B VanRenterghem et al.
The Journal of biological chemistry, 269(40), 24666-24672 (1994-10-07)
Previously pp60v-src, cyclin A, p39mos, and maturation-promoting factor (composed of Cdc2 and cyclin B) have been shown to activate mitogen-activated protein kinase (MAPK) and MAPK kinase (MEK) in cell-free extracts of Xenopus oocytes. The pp60v-src pathway is dependent on a
K P Huang et al.
Biochemical and biophysical research communications, 139(1), 320-326 (1986-08-29)
Tryptic fragments of protein kinase C containing the kinase (45 KDa) and phorbol ester-binding activity (38 KDa) were separated by Mono O column chromatography. The purified phorbol ester-binding fragment exhibits a higher affinity for phosphatidylserine than the native enzyme but
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service