P38 is a mitogen-activated protein kinase (MAPK) that belongs to serine/threonine protein kinase family and is a member of the stress-activated protein kinases (SAPK). P38 signaling pathway regulates inflammatory responses, apoptosis, cell proliferation and differentiation. Dually phosphorylated p38 (i.e (Thr180) and (Tyr182)) is a 43kD endogenous protein that belongs to High Osmolarity Glycerol response kinase (HOG). Anti-phospho-p38 (pThr180/pTyr182) antibody can be used in immunostaining and immunohistochemistry. Rabbit anti-phospho-p38 (pThr180/pTyr182) antibody reacts specifically with p38 of rat and human. This product is also reactive for endogenous, active forms of p38 α, β, γ (43kD).
Immunogen
synthetic phosphopeptide derived from the region of p38 that contains threonine180 and tyrosine182 based on the human sequence.
Application
Anti-phospho-p38 (pThr180/pTyr182) antibody can be used in immunofluorescence. It can also be used in western blotting (diluted 1: 1000).
Physical form
Solution in Dulbecco′s phosphate buffered saline (without Mg2+ and Ca2+), pH 7.3 with 50% glycerol, 1 mg/ml BSA and 0.05% sodium azide as a preservative.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
The Journal of experimental medicine, 216(5), 1182-1198 (2019-04-04)
Subclinical hypothyroidism is associated with cardiovascular diseases, yet the underlying mechanism remains largely unknown. Herein, in a common population (n = 1,103), TSH level was found to be independently correlated with both carotid plaque prevalence and intima-media thickness. Consistently, TSH
Recent evidence suggests that bone marrow (BM)-derived cells may integrate into the kidney, giving rise to functional renal cell types, including endothelial and epithelial cells and myofibroblasts. BM-derived cells can contribute to repair of the renal peritubular capillary (PTC) network
Age-related effects of the vascular wall have been associated with several hemodynamic dysfunctions, including medial vascular calcification. Vascular aging has been traditionally addressed using proliferative senescence of vascular smooth muscle cells (VSMC) in vitro, which induces osteoblastic transition and favors calcification
Cell biology and toxicology, 35(1), 67-80 (2018-07-08)
Aflatoxins are fungal metabolites classified into four major groups such as B1, B2, G1, and G2. These natural aflatoxins are designated as group I carcinogen by the International Agency for Research on Cancer. Among these, the aflatoxin B1 is more
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.