Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P1488

Sigma-Aldrich

Procaspase-3 human

>90% (SDS-PAGE), recombinant, expressed in E. coli

Synonym(s):

Apopain, CPP32, Yama

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200

recombinant

expressed in E. coli

assay

>90% (SDS-PAGE)

form

solid

mol wt

~30 kDa

solubility

PBS: soluble

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CASP3(836)

Biochem/physiol Actions

Can be cleaved and activated by caspase-8 and -10 and processed by caspase-9.

Other Notes

Contains 0.052% ammonium sulfate, 0.158% Tris-HCl, 0.76% sodium chloride.

Storage Class

13 - Non Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Besnault et al.
Journal of immunology (Baltimore, Md. : 1950), 167(2), 733-740 (2001-07-07)
We have previously reported that B cell receptors, depending on the degree to which they are cross-linked, can promote apoptosis in various human B cell types. In this study, we show that B cell receptors can trigger two apoptotic pathways

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service