Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P1369

Sigma-Aldrich

Anti-PMCA3 antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Plasma Membrane Calcium ATPase 3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat

concentration

~1 mg/mL

technique(s)

western blot: 1:1000 using rat tissue

UniProt accession no.

shipped in

wet ice

Storage temp.

−20°C

Gene Information

Immunogen

highly purified peptide corresponding to amino acid residues 5-22 of the rat PMCA3 protein.

Physical form

Solution in phosphate buffered saline containing 1 mg/mL bovine serum albumin and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anthony R D'Amato et al.
PloS one, 14(2), e0211731-e0211731 (2019-02-05)
Three aligned, electrospun fiber scaffolds with unique surface features were created from poly-L-lactic acid (PLLA). Fibers without surface nanotopography (smooth fibers), fibers with surface divots (shallow pits), and fibers with surface pits (deeper pits) were fabricated, and fiber alignment, diameter

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service