P1286
B-Phycoerythrin from Porphyridium cruentum
lyophilized powder
About This Item
Recommended Products
biological source
algae (Porphyridium cruentum)
form
lyophilized powder
composition
Protein, ~30% Lowry
concentration
20-40% protein (Lowry)
technique(s)
protein quantification: suitable
UniProt accession no.
storage temp.
2-8°C
Gene Information
Porphyridium cruentum ... cpeB(17963946)
Biochem/physiol Actions
Packaging
Physical form
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2
Storage Class
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service