Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P1248

Sigma-Aldrich

Anti-phospho-ATF-2 (pSer490/pSer498) antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

mol wt

~50 kDa

species reactivity

human

technique(s)

western blot: suitable using human melanoma cells incubated with varying doses of the radiomimetic drug NCS.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... ATF2(1386)

Immunogen

synthetic phosphopeptide corresponding to the region of human ATF2 containing serines 490 and 498.

Physical form

Solution in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service