Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P1168

Sigma-Aldrich

L-α-Phosphatidyl-D-myo-inositol 5-monophosphate, dipalmitoyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H80O16P2
CAS Number:
Molecular Weight:
891.01
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:

form

powder

solubility

DMSO: soluble (lit.)(lit.)
chloroform: soluble (lit.)(lit.)
ethanol: soluble (lit.)(lit.)

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCC(=O)OC[C@H](COP(O)(=O)O[C@@H]1[C@H](O)[C@H](O)[C@@H](O)[C@H](OP(O)(O)=O)[C@H]1O)OC(=O)CCCCCCCCCCCCCCC

InChI

1S/C41H80O16P2/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-34(42)53-31-33(55-35(43)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2)32-54-59(51,52)57-41-38(46)36(44)37(45)40(39(41)47)56-58(48,49)50/h33,36-41,44-47H,3-32H2,1-2H3,(H,51,52)(H2,48,49,50)/t33-,36-,37-,38-,39-,40+,41-/m1/s1

InChI key

SZPQTEWIRPXBTC-LNNNXZRXSA-N

Biochem/physiol Actions

Component of lipid signaling pathway.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hui Ma et al.
Cell research, 16(5), 466-478 (2006-05-16)
Multiple repeats of membrane occupation and recognition nexus (MORN) motifs were detected in plant phosphatidylinositl monophosphate kinase (PIPK), a key enzyme in PI-signaling pathway. Structural analysis indicates that all the MORN motifs (with varied numbers at ranges of 7-9), which
J E Harlan et al.
Biochemistry, 34(31), 9859-9864 (1995-08-08)
The pleckstrin homology (PH) domain is a protein module of approximately 100 amino acids that is found in several proteins involved in signal transduction [for a recent review, see Gibson et al. (1994) Trends Biochem. Sci. 19, 349-353]. Although the
C L Huang et al.
Nature, 391(6669), 803-806 (1998-03-05)
Inward rectifier K+ channels, which modulate electrical activity in many cell types, are regulated by protein kinases, guanine-nucleotide-binding proteins (G proteins) and probably actin cytoskeleton. Generation of phosphatidylinositol 4,5-bisphosphate (PIP2) by ATP-dependent lipid kinases is known to activate inward rectifier
J A Pitcher et al.
The Journal of biological chemistry, 270(20), 11707-11710 (1995-05-19)
The pleckstrin homology (PH) domain is an approximately 100-amino-acid region of sequence homology present in numerous proteins of diverse functions, which forms a discrete structural module. Several ligands capable of binding to PH domain-containing proteins have been identified including phosphatidylinositol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service