P0995
[pSer8]-Myelin Basic Protein Fragment 5-15 trifluoroacetate salt
≥90% (HPLC)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C57H96N23O18P
Molecular Weight:
1422.49
MDL number:
UNSPSC Code:
12352202
Recommended Products
biological source
synthetic (organic)
assay
≥90% (HPLC)
form
solid
storage temp.
−20°C
Amino Acid Sequence
Lys-Arg-Pro-pSer-Gln-Arg-His-Gly-Ser-Lys-Tyr-NH2
Application
Protein kinase C substrate
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service