Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P0959

Sigma-Aldrich

Gelatin solution

Protein standard, 2 mg/mL

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352204

Quality Level

concentration

2 mg/mL

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Packaging

Sealed ampules

Physical form

in 0.9% NaCl containing 0.05% sodium azide

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T S Anirudhan et al.
Journal of colloid and interface science, 456, 22-31 (2015-06-19)
A novel adsorbent, 2-mercaptobenzamide modified itaconic acid-grafted-magnetite nanocellulose composite [P(MB-IA)-g-MNCC] was synthesized for adsorbing mercury(II) [Hg(II)] ions selectively from aqueous solutions. Fourier transforms infrared spectroscopy, X-ray diffraction, scanning electron microscopy and thermogravimetric studies were performed to characterize the adsorbent. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service