Key Documents
P0867
Sigma-Aldrich
Hanna pHep pH meter - waterproof series
Model pHep 5
About This Item
UNSPSC Code:
41115603
manufacturer/tradename
Hanna HI98128
pH measuring range
0.00-14.00, accuracy: 0.05 pH unit, resolution: 0.01 pH unit
parameter
-5.0-60 °C temp. range
L × W × H
163 mm  mm × 26 mm
battery power
4 × 1.5 V, 350 hr life
General description
Pocket pHep waterproof pH tester with replaceable electrode cartridge.
  • Dual Level Display Showing pH & Temp
  • Percent Remaining Battery Indicator
  • Replaceable pH electrode featuring an extendable junction
  • Rugged, floating waterproof casing
  • Automatic calibration
  • Measurement stability indicator
  • The dual-level LCD displays: pH and °C or °F
  • Battery % displayed on start up with battery error prevention system (BEPS) to support optimum operation
Supplied with:
HI 73127 pH electrode, HI 73128 electrode removal tool, batteries and Instructions.
Choose from one of the most recent versions:
Certificates of Analysis (COA)