sterility
sterile-filtered
pH
4.2-4.9
capacity
0.35 mmol/pH·mL buffering capacity
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Other Notes
IEF carrier ampholytes
Legal Information
Pharmalyte is a registered trademark of Cytiva
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service