P0367
Hanna waterproof intelligent pH/ORP/ °C meter
HI 98150
About This Item
Recommended Products
manufacturer/tradename
Hanna
mV measuring range
±2000, accuracy: 0.4, resolution: 0.2
±600.0, accuracy: 0.2, resolution: 0.1
pH measuring range
-4.00-19.99, accuracy: 0.01 pH unit, resolution: 0.01 pH unit
temperature measuring range
-20.0-120 °C, accuracy: ±0.4 °C, resolution: 0.1
size
196 mm (7.7 in.) × 80 mm (3.1 in.) × 60 mm (2.4 in.)
weight
500 g
battery power
4 × 1.5 V (300 hours of continuous use)
General description
- IP 67 waterproof protection
- Automatic calibration data recall
- Dual level backlit LCD
- LCD guided calibration
- Password protected GLP data
- Supplied with electrode, DIN 1 M cable, 12 V DC adapter, pH 4.01/7.01 buffer solutions, cleaning solution, batteries, hard acrrying case and instruction manual.
- IP 67 waterproof protection
- Automatic calibration data recall
- Dual level backlit LCD
- LCD guided calibration
- Password protected GLP data
- Supplied with electrode, DIN 1 M cable, 12 V DC adapter, pH 4.01/7.01 buffer solutions, cleaning solution, batteries, hard acrrying case and instruction manual
Hints on Electrode Maintenance
Choose from one of the most recent versions:
Certificates of Analysis (COA)