Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P0247

Sigma-Aldrich

L-NASPA

≥98% (TLC), powder

Synonym(s):

N-Palmitoyl-L-serine phosphoric acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H38NO7P
CAS Number:
Molecular Weight:
423.48
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

assay

≥98% (TLC)

form

powder

color

off-white

solubility

DMSO: 10 mg/mL
ethanol: 10 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCC(=O)N[C@@H](COP(O)(O)=O)C(O)=O

InChI

1S/C19H38NO7P/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-18(21)20-17(19(22)23)16-27-28(24,25)26/h17H,2-16H2,1H3,(H,20,21)(H,22,23)(H2,24,25,26)/t17-/m0/s1

InChI key

HSMUPNYJCMYKJH-KRWDZBQOSA-N

Application

Used to characterize the LPA receptor (Edg-7). EC50 = 5.3 nM.

Biochem/physiol Actions

Competitive mammalian lysophosphatitic acid (LPA) receptor agonist; LPA mimetic; potent reversible antagonist of the LPA receptors expressed in Xenopus oocytes.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service