Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P0243

Sigma-Aldrich

Phalloidin, poly-L-Lysine bound

≥90%

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200

assay

≥90%

form

solid

mol wt

average mol wt ~25 kDa

solubility

H2O: soluble

shipped in

wet ice

storage temp.

2-8°C

General description

By weight, each vial contains 20% phalloidin (200 μg), 50% poly-L-lysine with the balance as sodium chloride as a stabilizer.

Biochem/physiol Actions

It is predicted that poly-L-lysine bound phalloidin is approximately 100 fold more potent than the unbound form.
Toxin that binds polymeric F actin, stabilizing it and interfering with the function of actin-rich structures.

Caution

Highly toxic

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral

Storage Class

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service