P0240
PRAK Substrate Peptide
1 mg/mL in H2O, ≥98% (HPLC), aqueous solution
Synonym(s):
P38 Regulated/Activated Protein Kinase Substrate Peptide
About This Item
Quality Level
assay
≥98% (HPLC)
form
aqueous solution
mol wt
1171 by amino acid sequence
concentration
1 mg/mL in H2O
shipped in
dry ice
storage temp.
−20°C
SMILES string
CC(C)C[C@H](NC(=O)[C@H](CCCCN)NC(=O)[C@@H](N)CCCCN)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CO)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](C)C(O)=O
InChI
1S/C51H98N18O13/c1-26(2)23-35(65-42(74)32(16-10-12-20-53)62-40(72)31(54)15-9-11-19-52)44(76)64-33(17-13-21-59-50(55)56)41(73)63-34(18-14-22-60-51(57)58)43(75)69-39(30(8)71)48(80)66-36(24-27(3)4)45(77)67-37(25-70)46(78)68-38(28(5)6)47(79)61-29(7)49(81)82/h26-39,70-71H,9-25,52-54H2,1-8H3,(H,61,79)(H,62,72)(H,63,73)(H,64,76)(H,65,74)(H,66,80)(H,67,77)(H,68,78)(H,69,75)(H,81,82)(H4,55,56,59)(H4,57,58,60)/t29-,30+,31-,32-,33-,34-,35-,36-,37-,38-,39-/m0/s1
InChI key
QDSZECPRZNYZJS-DVHCFKBKSA-N
Amino Acid Sequence
Biochem/physiol Actions
Features and Benefits
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service