Key Documents
P0185
Pipette tips, 200 μL
With aerosol barrier, usable capacity 140μL, sterile
About This Item
UNSPSC Code:
41121601
NACRES:
NB.01
material
colorless
sterility
sterile
packaging
pack of 96 ea (Trays)
maximum volume
200 μL
General description
Pipette tips (200μL) contain inert, bonded fiber barrier which coalesces airborne contaminants so they do not enter pipette barrel. Placement of barrier reduces usable capacity to 140μL. These are same tips as P 9934, but barrier placed higher up, allowing greater capacity; usable capacity is 140μL.
Other Notes
Browse all pipette tips
Choose from one of the most recent versions:
Certificates of Analysis (COA)