Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

P0120

Sigma-Aldrich

Anti-Purinergic Receptor P2Y13 antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-P2Y Purinoceptor 13

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

rat

technique(s)

western blot: 1:200 using rat brain membranes

UniProt accession no.

shipped in

dry ice

Storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Immunogen

peptide DRFLKIIRPLRNIFLK(C) corresponding to residues 119-134 of human P2Y13 receptor.

Target description

P2Y13 receptor belongs to the G-protein coupled receptors superfamily. Like the P2Y12 receptor it is a high affinity receptor for the ADP nucleotide, coupled to the Gai class of G-proteins. The P2Y13 receptor is highly expressed at brain and spleen tissues as well as in the immune system. ADP activates a negative feedback pathway for ATP release from human red blood cells via the P2Y13 receptor. P2Y13 receptor was also implicated in inhibition of N-type Ca2+ channels in neurons, exerting pre and/or post-synaptic modulatory action.

Physical form

Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA and 0.05% sodium azide.

Reconstitution

Reconstitute the lyophilized vial with 50 μL or 200 μL deionized water, depending on package size. Further dilutions should be made using a carrier protein such as BSA (1-3%).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ov D Slayden et al.
Journal of medical primatology, 52(6), 405-413 (2023-10-18)
Endometriosis is the presence of endometrium-like tissue outside the uterine cavity. An experimental model of endometriosis has been created in the baboon by the transcervical collection and laparoscopic inoculation of menstrual endometrium. Macaques are the preferred model for pharmaceutical development

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service