Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

P0118

Sigma-Aldrich

Anti-Potassium Channel Kv4.1 antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-KCND1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human, rat, mouse

technique(s)

western blot: 1:400 using human neuroblastoma cell line SH-SY5Y

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KCND1(3750)
mouse ... Kcnd1(16506)
rat ... Kcnd1(116695)

Immunogen

peptide (C)KRRAIRLANSTAS, corresponding to residues 538-550 of the human Kv4.1.The epitope is localized intracellularly in C-terminal.

Target description

The Kv4 channels are characterized by activation at subthreshold membrane potentials, inactivate rapidly and recover from inactivation quickly compared with other voltage-dependent K+ channels. This type of current is known as transient A-type K+ currents. For example, depolarization-activated K+ currents in rat neostriatal cholinergic interneurons are predominantly of the A-type and attributable to coexpression of Kv4.1 and Kv4.2 subunits.

Physical form

Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA and 0.025% sodium azide.

Reconstitution

Reconstitute the lyophilized vial with 50 μL or 200 μL deionized water, depending on package size. Further dilutions should be made using a carrier protein such as BSA (1-3%).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elif Sarinay Cenik et al.
Developmental cell, 48(6), 811-826 (2019-02-26)
Caenorhabditis elegans provides an amenable system to explore whether newly composed ribosomes are required to progress through development. Despite the complex pattern of tissues that are formed during embryonic development, we found that null homozygotes lacking any of the five

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service