Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

P0096

Sigma-Aldrich

Anti-POT1 antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-POT1-like telomere end-binding protein, Anti-PROTECTION OF TELOMERES 1, Anti-hPOT1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 55-58 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

technique(s)

western blot: 1:1,000-1:2,000 using lysate of C2 or HeLa nuclear extract

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... POT1(25913)

General description

POT1 (Protection of Telomere 1, also known as POTE1 and POT1-like telomere end-binding protein) is a highly conserved single-strand telomeric binding protein. POT1 is one of the six core telomere-binding proteins termed the telosome2 or shelterin. POT1 has eukaryotic homologs and first identified in S. pombe. Mice that carry two POT1 genes encoding two proteins (POT1a and POT1b). Human genome contains one gene that encodes four alternative splice variants (v2, v3, v4, and v5).

Application

Anti-POT1 antibody produced in rabbit has been used in immunoblotting.

Biochem/physiol Actions

Protection of Telomere 1 (POT1) binds single-stranded telomeric DNA through two NH2-terminal oligonucleotide-binding folds and interacts with TTP1 through the COOH-terminal region. It is required for both chromosomal end protection and telomere length regulation. Genetic disruption of the two POT1 genes in mice revealed two different phenotypes. Knockout of Pot1a alone results in embryonic lethality. Pot1a/b double knockout cells showed enhanced telomere dysfunction induced foci.

Target description

POT1 (Protection of Telomere 1, also known as POTE1and POT1-like telomere end-binding protein) is a highly conserved single-strand telomeric binding protein and is required for both chromosomal end protection and telomere length regulation.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Recent expansion of the telomeric complex in rodents: Two distinct POT1 proteins protect mouse telomeres
Hockemeyer D, et al.
Cell, 126(1), 63-77 (2006)
Functional diversity of human protection of telomeres 1 isoforms in telomere protection and cellular senescence
Yang Q, et al.
Cancer Research, 67(24), 11677-11686 (2007)
Pot1 deficiency initiates DNA damage checkpoint activation and aberrant homologous recombination at telomeres
Wu L, et al.
Cell, 126(1), 49-62 (2006)
Cory Rice et al.
Nature communications, 8, 14928-14928 (2017-04-11)
POT1 and TPP1 are part of the shelterin complex and are essential for telomere length regulation and maintenance. Naturally occurring mutations of the telomeric POT1-TPP1 complex are implicated in familial glioma, melanoma and chronic lymphocytic leukaemia. Here we report the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service