Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P0070

Sigma-Aldrich

Anti-Presenilin-2, N-terminal antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-AD3L, Anti-AD4, Anti-PS2, Anti-PSEN2, Anti-Presenilin 2 (Alzheimer 4), Anti-STM2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~50 kDa

species reactivity

human

technique(s)

indirect ELISA: 1:1,000
western blot: 1:100-1:500

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... PSEN2(5664)

Looking for similar products? Visit Product Comparison Guide

Immunogen

synthetic peptide corresponding to the N-terminal of human presenilin-2.

Physical form

Solution in phosphate buffered saline with 0.09% (w/v) sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xike Qin et al.
The Journal of biological chemistry, 291(42), 22276-22287 (2016-09-01)
Accumulation of amyloid-β peptide (Aβ) in the brain is regarded as central to Alzheimer's disease (AD) pathogenesis. Aβ is generated by a sequential cleavage of amyloid precursor protein (APP) by β-secretase 1 (BACE-1) followed by γ-secretase. BACE-1 cleavage of APP

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service