Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P0028

Sigma-Aldrich

PSN-375963

≥98% (HPLC)

Synonym(s):

4-[5-(4-butylcyclohexyl)-1,2,4-oxadiazol-3-yl]-pyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H23N3O
CAS Number:
Molecular Weight:
285.38
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: >10 mg/mL

storage temp.

−20°C

SMILES string

CCCCC1CCC(CC1)c2nc(no2)-c3ccncc3

InChI

1S/C17H23N3O/c1-2-3-4-13-5-7-15(8-6-13)17-19-16(20-21-17)14-9-11-18-12-10-14/h9-13,15H,2-8H2,1H3

InChI key

OAVLEYPTWABFLF-UHFFFAOYSA-N

Biochem/physiol Actions

PSN-375963 is a GPR119 partial agonist.
PSN-375963 is a GPR119 partial agonist. GPR119 is an orphan GPCR, expressed predominantly in pancreas and GI tract, that has been characterized. Its endogenous ligands are oleoylethanolamide (OEA) and oleoyl-lysophosphatidylcholine, lipid signaling molecules that regulate food intake and body weight. OEA decreases food intake, so GPR119 agonists are potential therapies for obesity. Initial studies showed that PSN-375963 has similar potency and partial efficacy compared to OEA. Further studies revealed that although OEA and PSN-375963 are GPR119 agonists, they have divergent effects on insulin secretion, calcium signaling, and cAMP production.

Storage Class

11 - Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Madusha Peiris et al.
Nutrients, 10(10) (2018-10-20)
Nutrient-sensing receptors located on enteroendocrine (EEC) cells modulate appetite via detection of luminal contents. Colonic 'tasting' of luminal contents may influence changes to appetite observed in obesity and after weight loss induced by bariatric surgery. We assessed the effects of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service