Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

OGS411

Sigma-Aldrich

PSF-CMV-CMV-SBFI - DUAL CMV EXPRESSION PLASMID

plasmid vector for molecular cloning

Synonym(s):

cloning vector, expression vector, molecular cloning vector, plasmid, plasmid vector, snapfast vector, vector

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.85
Product OGS411 is not currently sold in your country. Contact Technical Service

form

buffered aqueous solution

mol wt

size 5131 bp

bacteria selection

kanamycin

origin of replication

pUC (500 copies)

peptide cleavage

no cleavage

promoter

Promoter name: CMV
Promoter activity: constitutive
Promoter type: mammalian

reporter gene

none

shipped in

ambient

storage temp.

−20°C

General description

PSF-CMV-CMV-SBFI - dual CMV expression plasmid is a versatile expression vector for the production of two proteins under the control of two separate CMV promoter expression cassettes in mammalian cells. This vector also contains a Kanamycin resistance cassette for growth and maintenance in E. coli.

Promoter Expression Level: This plasmid vector contains the mammalian CMV promoter to drive gene expression. We have tested all of our mammalian promoters in a range of cell types and CMV is consistently the strongest in those we have studied. However there are many reports of the CMV promoter demonstrating silencing by methylation in long-term culture.

Application

First multiple cloning site notes: There is start codon in the NcoI site that can be removed by digestion with KpnI if required. The first MCS for gene insertions extends from NotI to XbaI but the Shine-Dalgarno sequences and Kozak sequences are aligned with the start codon in the NcoI restriction site. The ClaI to NheI sites have other functions such as adding peptide tags or IRES elements. The BsgI and BseRI restriction sites cleave within the stop codon in the XbaI site and allow the retrospective fusion of coding sequences. These sites are normally only used on genes that we sell in the main multiple cloning site.

Second multiple cloning site notes: The second MCS in PSF-CMV-CMV-SBFI - dual CMV expression plasmid extends from PspOMI to SpeI. This MCS contains a Kozak sequence immediately upstream of the start codon in the PciI restriction site. We insert the start codons of our genes into here where possible.

This MCS has been designed to be compatible with genes that are within our main MCS by using enzymes sites in the same order that produce compatible cohesive ends that can be ligated together. This allows genes to be transferred from the main MCS (NotI to NheI) into the second MCS (PspOMI to SpeI) is required. Enzymes that are compatible between these two MCSs include:

  • PspOMI compatible with NotI
  • ScaI compatible with EcoRV (Blunt)
  • SalI compatible with XhoI
  • PciI compatible with NcoI and BspHI
  • AclI compatible with ClaI
  • SpeI compatible with XbaI and NheI Avr2

Sequence

To view sequence information for this product, please visit the product page

Analysis Note

To view the Certificate of Analysis for this product, please visit www.oxgene.com

related product

Product No.
Description
Pricing

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diana Romero et al.
Carcinogenesis, 37(1), 18-29 (2015-10-28)
Dickkopf-3 (Dkk-3) is a secreted protein whose expression is downregulated in many types of cancer. Endogenous Dkk-3 is required for formation of acini in 3D cultures of prostate epithelial cells, where it inhibits transforming growth factor (TGF)-β/Smad signaling. Here, we
Geoffrey M Lynn et al.
Nature biotechnology, 33(11), 1201-1210 (2015-10-27)
The efficacy of vaccine adjuvants such as Toll-like receptor agonists (TLRa) can be improved through formulation and delivery approaches. Here, we attached small molecule TLR-7/8a to polymer scaffolds (polymer-TLR-7/8a) and evaluated how different physicochemical properties of the TLR-7/8a and polymer
Jin-Gyoung Jung et al.
PLoS genetics, 10(10), e1004751-e1004751 (2014-10-31)
The Notch3 signaling pathway is thought to play a critical role in cancer development, as evidenced by the Notch3 amplification and rearrangement observed in human cancers. However, the molecular mechanism by which Notch3 signaling contributes to tumorigenesis is largely unknown.
Alexander C Cerny et al.
PLoS genetics, 11(10), e1005578-e1005578 (2015-10-29)
Recycling of signaling proteins is a common phenomenon in diverse signaling pathways. In photoreceptors of Drosophila, light absorption by rhodopsin triggers a phospholipase Cβ-mediated opening of the ion channels transient receptor potential (TRP) and TRP-like (TRPL) and generates the visual

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service