OGS185
PSF-T7-EMCV - T7 IRES EXPRESSION PLASMID
plasmid vector for molecular cloning
Synonym(s):
cloning vector, expression vector, molecular cloning vector, plasmid, plasmid vector, snapfast vector, vector
About This Item
Recommended Products
form
buffered aqueous solution
mol wt
size 4378 bp
bacteria selection
kanamycin
origin of replication
pUC (500 copies)
peptide cleavage
no cleavage
promoter
Promoter name: T7
Promoter activity: inducible
Promoter type: phage
reporter gene
none
shipped in
ambient
storage temp.
−20°C
General description
application
The ClaI to NheI sites have other functions such as adding peptide tags second promoters or other IRES elements. The BsgI and BseRI restriction sites cleave within the stop codon in the XbaI site and allow the retrospective fusion of coding sequences. These sites are normally only used on genes that we sell in the main multiple cloning site.
Sequence
Analysis Note
related product
Storage Class
12 - Non Combustible Liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service