Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

OGS1200

Sigma-Aldrich

PSF-CMV-PURO-COOH-THR-FLAG®-6HIS - C-TERMINAL 6 HIS AND FLAG® DUAL TAG MAMMALIAN PLASMID

plasmid vector for molecular cloning

Synonym(s):

cloning vector, expression vector, molecular cloning vector, plasmid, plasmid vector, snapfast vector, vector

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.85

tag

6-His tagged
FLAG® tagged

form

buffered aqueous solution

mol wt

size 6210 bp

bacteria selection

kanamycin

mammalian cells selection

puromycin

origin of replication

pUC (500 copies)

peptide cleavage

thrombin

peptide tag location

C-terminal

promoter

Promoter name: CMV
Promoter activity: constitutive
Promoter type: mammalian

reporter gene

none

shipped in

ambient

storage temp.

−20°C

General description

This plasmid is designed to express tagged proteins in mammalian cells either by transient transfection or by creating stable cell lines. It contains a puromycin resistance expression cassette using the human Ubiquitin promoter to drive expression and allow for the selection of cells containing the plasmid.

About the Peptide Tag:This plasmid contains a c-terminal FLAG epitope tag that can be fused to a gene of interest to allow protein detection and/or purification. The sequence of the tag is: DYKDDDDK. This plasmid also contains a secondary Hexa-Histidine (6His) tag protein tag. The sequence of this tag is: HHHHHHWe provide a range of dual peptide tag plasmids. This is because some peptide tags provide specific biological properties (e. g., small molecule affinity new epitopes solubility or protein secretion) that are not provided by others.

About the Cleavage Tag: This plasmid also encodes a protease cleavage site that is designed to be positioned between your gene of interest and the tag to allow the removal of the tag following protein purification or isolation. This plasmid contains a Thrombin cleavage tag. The protein sequence of the cleavage tag is: LVPRGS. It cleaves preferentially between the Arg and Gly residues. Off target cleavage can often occur at non-specific sites normally from other contaminating proteases. To ensure maximal protein integrity the enzyme reagent must be highly pure.

Promoter Expression Level: This plasmid contains the mammalian CMV promoter to drive gene expression. We have tested all of our mammalian promoters in a range of cell types and CMV is consistently the strongest in those we have studied. However there are many reports of the CMV promoter demonstrating silencing by methylation in long-term culture.

Sequence

Please select the file type you require. For reference most cloning programs will import a .gb (Genbank) file and will show all of the plasmids features automatically when downloaded and imported.

Genebank Vector Sequence File

FASTA Vector Sequence File

Full Plasmid Map

Analysis Note

To view the Certificate of Analysis for this product, please visit www.oxfordgenetics.com.

Other Notes

Looking for more vector options to move your experiments forward faster? Consider a custom cloning vector designed and built by Oxford Genetics. Find out more at Oxford Genetics - Sigma's partner for cloning and expression vectors for molecular biology and synthetic biology applications.

Legal Information

FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
Oxford Genetics is a trademark of Oxford Genetics Ltd

related product

Product No.
Description
Pricing

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tomomi Yamamoto-Fukuda et al.
European archives of oto-rhino-laryngology : official journal of the European Federation of Oto-Rhino-Laryngological Societies (EUFOS) : affiliated with the German Society for Oto-Rhino-Laryngology - Head and Neck Surgery, 272(10), 2689-2696 (2014-08-21)
We reported previously that keratinocyte growth factor (KGF), a mesenchymal cell-derived paracrine growth factor, plays an important role in middle ear cholesteatoma formation, which is characterized by marked proliferation of epithelial cells. Here, we investigated whether KGF, the main factor
Jin-Gyoung Jung et al.
PLoS genetics, 10(10), e1004751-e1004751 (2014-10-31)
The Notch3 signaling pathway is thought to play a critical role in cancer development, as evidenced by the Notch3 amplification and rearrangement observed in human cancers. However, the molecular mechanism by which Notch3 signaling contributes to tumorigenesis is largely unknown.
Alexander C Cerny et al.
PLoS genetics, 11(10), e1005578-e1005578 (2015-10-29)
Recycling of signaling proteins is a common phenomenon in diverse signaling pathways. In photoreceptors of Drosophila, light absorption by rhodopsin triggers a phospholipase Cβ-mediated opening of the ion channels transient receptor potential (TRP) and TRP-like (TRPL) and generates the visual
Geoffrey M Lynn et al.
Nature biotechnology, 33(11), 1201-1210 (2015-10-27)
The efficacy of vaccine adjuvants such as Toll-like receptor agonists (TLRa) can be improved through formulation and delivery approaches. Here, we attached small molecule TLR-7/8a to polymer scaffolds (polymer-TLR-7/8a) and evaluated how different physicochemical properties of the TLR-7/8a and polymer
Diana Romero et al.
Carcinogenesis, 37(1), 18-29 (2015-10-28)
Dickkopf-3 (Dkk-3) is a secreted protein whose expression is downregulated in many types of cancer. Endogenous Dkk-3 is required for formation of acini in 3D cultures of prostate epithelial cells, where it inhibits transforming growth factor (TGF)-β/Smad signaling. Here, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service