Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

OGR007

Sigma-Aldrich

SnapFast Master Mix

(Cloning Kit for SnapFast Vectors)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

shelf life

Avoid repeated freeze/thaw cycles

packaging

pkg of 1 vial (OGR007-1EA)
pkg of 10 vials (OGR007X10-1EA)
pkg of 5 vials (OGR007X5-1EA)

solvent strength

2x (Supplied as 2x concentrate in buffer)

shipped in

wet ice

storage temp.

−20°C

General description

SnapFastTM DNA Assembly Master Mix is a high performance single tube DNA assembly enzyme mix that allows the efficient joining of any number of DNA molecules without restriction enzymes. DNA molecules with suitable homology (20 bp or more) can be joined efficiently in a single step reaction, and several molecules can be joined simultaneously to make long DNA molecules. SnapFast Master Mix is suitable for use with both long and short DNA sequences, and mixtures of long and short DNA molecules can be joined simultaneously. Click here to view the protocol.

Other Notes

Vendor Tech Service Contact information (phone # ): +44 (0) 1865 309 664

Vendor Tech Service Contact information (email): [email protected]

Legal Information

SnapFast by Oxford Genetics

Oxford Genetics is a trademark of Oxford Genetics Ltd

related product

Product No.
Description
Pricing

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service