Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

O9887

Sigma-Aldrich

Oleyl trifluoromethyl ketone

≥98%, ethanol solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H33F3O
CAS Number:
Molecular Weight:
334.46
EC Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

biological source

synthetic (organic)

Quality Level

assay

≥98%

form

ethanol solution

shipped in

wet ice

storage temp.

−20°C

Biochem/physiol Actions

Inhibitor of fatty acid amide hydrolase.

pictograms

FlameExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

57.2 °F - closed cup

flash_point_c

14 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service