Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

O8762

Sigma-Aldrich

Oncostatin M Receptor β/Fc Chimera from mouse

>90% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder

Synonym(s):

OSM Rβ

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352204

recombinant

expressed in NSO cells

assay

>90% (SDS-PAGE)

form

lyophilized powder

mol wt

~150 kDa by SDS-PAGE (reducing)
calculated mol wt 109 kDa

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

Biochem/physiol Actions

Exclusive signal transducing receptor protein for mouse OSM. Mouse and human OSM Rβ share 55% sequence identity.

Other Notes

Extracellular domain of mouse OSM Rβ (a.a. 1-738) fused to the C-terminal 6X histidine-tagged Fc region of human IgG1.

Physical form

Lyophilized from 0.2 μm filtered solution in phosphate buffered saline.

Analysis Note

The biological activity is measured by its ability to bind recombinant mouse OSM.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service