Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

O6762

Sigma-Aldrich

BioParticles® Opsonizing Reagents

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151817
NACRES:
NA.56

compatibility

for use with S. aureus Wood strain

Quality Level

storage temp.

−20°C

Application

Opsonizing reagents are used in affinity chromatography and protein chromatography. Opsonizing reagents have been used in studies to develop an easy and inexpensive method of investigating phagocytosis in retinal pigment epithelium cells. They have also been used to study roles in innate defense against infection and to investigate immunomodulating activity of medicinal plants.

Legal Information

BioParticles is a registered trademark of Life Technologies

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W S Koko et al.
Journal of ethnopharmacology, 118(1), 26-34 (2008-04-29)
Ethanolic extracts of 23 medicinal plants, commonly used in Sudanese folk medicines against infectious diseases, were investigated for their immunomodulating activity using luminol/lucigenin-based chemiluminescence assay. Preliminary screenings on whole blood oxidative burst activity showed inhibitory activities of 14 plant extracts
Alexa Klettner et al.
Ophthalmic research, 46(2), 88-91 (2011-01-29)
The retinal pigment epithelium (RPE) is the most active phagocyte in the human body, and this function is indispensable for vision. In this study, we introduce an easy and inexpensive way to quantify RPE phagocytosis by opsonizing FITC-labeled latex beads
Viral aggregating and opsonizing activity in collectin trimers
Hartshorn, K.L., et al.
American Journal of Physiology. Lung Cellular and Molecular Physiology, 298, 79-88 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service