Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

O4514

Sigma-Aldrich

Anti-Orexin Receptor 1 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-OX1R

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

availability

not available in Japan

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 14 μg/mL using human brain tissue

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... HCRTR1(3061)
mouse ... Hcrtr1(230777)
rat ... Hcrtr1(25593)

Immunogen

OX1R / Orexin Receptor 1 antibody was raised against synthetic 16 amino acid peptide from C-terminus of human HCRTR1 / Orexin Receptor 1

Biochem/physiol Actions

Orexin is a neuropeptide that modulates the energy and metabolism balance. The effects of orexin are mediated by one of the two G protein-coupled receptors, orexin receptor 1 or 2. OXRs are expressed in oxinergic projection areas of the brain and regulate food intake by reducing systemic and brain signals that drive over-eating.

Physical form

Solution in phosphate buffered saline containing 0.1% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas E Fitch et al.
Frontiers in neuroscience, 8, 5-5 (2014-01-31)
We describe a novel, potent and selective orexin-2 (OX2)/hypocretin-2 receptor antagonist with in vivo activity in an animal model predictive of antidepressant-like efficacy. N-biphenyl-2-yl-4-fluoro-N-(1H-imidazol-2-ylmethyl) benzenesulfonamide HCl (LSN2424100) binds with high affinity to recombinant human OX2 receptors (Ki = 4.5 nM)
Yonghua Chen et al.
Frontiers in cellular neuroscience, 13, 478-478 (2019-11-12)
Airway vagal nerves play a predominant role in the neural control of the airway, and augmented airway vagal activity is known to play important roles in the pathogenesis of some chronic inflammatory airway diseases. Several lines of evidence indicate that
Michel A Steiner et al.
Frontiers in pharmacology, 4, 165-165 (2014-01-15)
The orexin system regulates feeding, nutrient metabolism and energy homeostasis. Acute pharmacological blockade of orexin receptor 1 (OXR-1) in rodents induces satiety and reduces normal and palatable food intake. Genetic OXR-1 deletion in mice improves hyperglycemia under high-fat (HF) diet
Zi-Bing Liu et al.
Clinical and experimental pharmacology & physiology, 37(10), 963-968 (2010-06-10)
1. Orexins are neuropeptides synthesized in the hypothalamus that regulate many physiological functions, including energy homeostasis, stress responses, sleep/wake states etc. It is now emerging that orexins may also regulate breathing, but little is known as to how they do

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service