Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

O2757

Sigma-Aldrich

3-Oxo-4-aza-5α-androstane-17β-(N-t-butylcarboxamide)

≥90%, powder

Synonym(s):

Dihydroproscar

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$231.00

$231.00


Available to ship onMay 02, 2025Details


Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
5 MG
$231.00

About This Item

Empirical Formula (Hill Notation):
C23H38N2O2
CAS Number:
Molecular Weight:
374.56
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77
form:
powder
assay:
≥90%

$231.00


Available to ship onMay 02, 2025Details


Notify Me

Get notified when this item is ready to ship via email.

assay

≥90%

form

powder

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

color

yellow

solubility

chloroform: 50 mg/mL, clear, colorless to yellow

shipped in

ambient

storage temp.

−20°C

SMILES string

[H][C@@]12CC[C@@]3([H])[C@]4([H])CC[C@H](C(=O)NC(C)(C)C)[C@@]4(C)CC[C@]3([H])[C@@]1(C)CCC(=O)N2

InChI

1S/C23H38N2O2/c1-21(2,3)25-20(27)17-8-7-15-14-6-9-18-23(5,13-11-19(26)24-18)16(14)10-12-22(15,17)4/h14-18H,6-13H2,1-5H3,(H,24,26)(H,25,27)/t14-,15-,16-,17+,18+,22-,23+/m0/s1

InChI key

ZOIUUCNFVDJSJK-WSBQPABSSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service