Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

O2264

Sigma-Aldrich

ONO-1301

≥98% (HPLC)

Synonym(s):

7,8-Dihydro-5-[(E)-[[α-(3-pyridyl)benzylidene]aminooxy]ethyl]-1-naphthyloxy]acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H24N2O4
CAS Number:
Molecular Weight:
428.48
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥98% (HPLC)

composition

Mixture of E and Z isomers

color

white

mp

147-158 °C (lit.)

solubility

DMSO: 18 mg/mL

storage temp.

−20°C

SMILES string

OC(=O)COc1cccc2C(CCO\N=C(/c3ccccc3)c4cccnc4)=CCCc12

InChI

1S/C26H24N2O4/c29-25(30)18-31-24-13-5-11-22-19(9-4-12-23(22)24)14-16-32-28-26(20-7-2-1-3-8-20)21-10-6-15-27-17-21/h1-3,5-11,13,15,17H,4,12,14,16,18H2,(H,29,30)/b28-26+

InChI key

WBBLIRPKRKYMTD-BYCLXTJYSA-N

Gene Information

Biochem/physiol Actions

Non-prostanoid prostacyclin mimetic; IP receptor agonist with weak EP3 agonist activity.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiroaki Obata et al.
American journal of respiratory and critical care medicine, 177(2), 195-201 (2007-11-03)
Although prostacyclin is recognized as a therapeutic breakthrough for pulmonary hypertension, it needs continuous infusion because of its short action. Therefore, we developed a new drug delivery system for prostacyclin. We prepared ONO-1301MS, a novel sustained-release prostacyclin analog polymerized with
Hiroshi Iwata et al.
Life sciences, 85(5-6), 255-261 (2009-06-17)
It was reported that administration of angiogenic growth factors can augment collateral growth in ischemic tissues. It is assumed that angiogenic effects of cell transplantation may be mainly mediated by secretion of angiogenic cytokines. We tested feasibility of clinical use
Qing Xu et al.
American journal of physiology. Gastrointestinal and liver physiology, 302(4), G420-G429 (2011-12-14)
Previous studies have demonstrated that mice disrupted with the cyclooxygenase-2 gene showed much more severe liver damage compared with wild-type mice after liver injury, and prostaglandins (PGs) such as PGE(1/2) and PGI(2) have decreased hepatic injury, but the mechanisms by
Yoichiro Hirata et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 63(10), 781-786 (2009-11-13)
Impairment of cardiac function in cardiomyopathy has been postulated to be related to decreased blood flow and increased collagen synthesis. Administration of growth factors was reported to attenuate left ventricular (LV) remodeling and dysfunction in animal models of dilated cardiomyopathy.
Cristiane Yamabayashi et al.
American journal of respiratory cell and molecular biology, 47(2), 170-177 (2012-03-10)
Airway remodeling in bronchial asthma results from chronic, persistent airway inflammation. The effects of the reversal of airway remodeling by drug interventions remain to be elucidated. We investigated the effects of ONO-1301, a novel prostacyclin agonist with thromboxane inhibitory activity

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service