Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

O0516

Sigma-Aldrich

Oncrasin-1

≥98% (HPLC)

Synonym(s):

1-[(4-Chlorophenyl)methyl]-1H-indole-3-carboxaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H12ClNO
CAS Number:
Molecular Weight:
269.73
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

off-white to light tan

solubility

DMSO: ≥20 mg/mL

storage temp.

2-8°C

SMILES string

Clc1ccc(Cn2cc(C=O)c3ccccc23)cc1

InChI

1S/C16H12ClNO/c17-14-7-5-12(6-8-14)9-18-10-13(11-19)15-3-1-2-4-16(15)18/h1-8,10-11H,9H2

InChI key

ZDRQMXCSSAPUMM-UHFFFAOYSA-N

Biochem/physiol Actions

Oncrasin-1 is a suppressor of RNA processing machinery. Oncrasin-1 induces aggregation of PKCL in the nuclei and is proapoptotic.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service