N8539
Anti-NDRG1 (C-terminal) antibody produced in rabbit
~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution
Synonym(s):
Anti-CAP43, Anti-CMT4D, Anti-DRG1, Anti-GC4, Anti-HMSNL, Anti-N-myc downstream regulated 1, Anti-NDR1 (differentiation-related gene 1 protein), Anti-NMSL, Anti-PROXY1 (protein regulated by oxygen 1), Anti-RIT42, Anti-RTP (reducing agents and tunicamycin-responsive protein), Anti-TARG1, TDD5
About This Item
biological source
rabbit
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
antigen ~43 kDa
species reactivity
mouse, human, rat
packaging
antibody small pack of 25 μL
concentration
~1.0 mg/mL
technique(s)
immunoprecipitation (IP): 5-10 μg using lysates of HeLa cells
western blot: 1-2 μg/mL using whole extract of mouse 3T3 and rat NRK cells
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... NDRG1(10397)
General description
Application
Biochem/physiol Actions
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service