Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N8395

Millipore

Filter membranes, nitrocellulose

MF membrane, pore size 0.22 μm, diam. 13 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104900
PubChem Substance ID:
NACRES:
NB.24

material

MF membrane

manufacturer/tradename

Millipore GSWP01300

diam.

13 mm

pore size

0.22 μm

Looking for similar products? Visit Product Comparison Guide

General description

Biologically inert nitrocellulose membranes containing a small amount of cellulose acetate for improved handling. Autoclavable to 121°C; use below 75°C. Chemically compatible with dilute acids and bases, hydrocarbons, and non-polar liquids. MF-Millipore membranes have water extractables <5%, including a Triton wetting agent TF-Millipore membranes are Triton-free and have even lower water extractables than MF membranes. See Immobilon-NC in the Electrophoresis Equipment section for nitrocellulose blotting membranes.

pictograms

Flame

signalword

Danger

hcodes

pcodes

Hazard Classifications

Flam. Sol. 1

Storage Class

4.1B - Flammable solid hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dezhi Chen et al.
Molecular medicine reports, 23(2) (2020-12-15)
Diabetic cardiomyopathy (DCM) is one of the primary complications of the cardiovascular system due to diabetes‑induced metabolic injury. The present study investigated the autophagy‑associated regulatory mechanisms of long non‑coding RNAs in cardiac pathological changes in diabetes mellitus (DM). Streptozotocin (STZ)‑induced
Dinesh Chandra et al.
Clinical and experimental immunology, 154(2), 224-234 (2008-09-10)
The failure of Leishmania, an intracellular pathogen, to stimulate a pro-inflammatory response following entry into macrophages has been well reported. This occurs in spite of the fact that ligands for the toll-like receptors (TLR) have been recently shown on the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service