Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

N8278

Sigma-Aldrich

N-(NBD-Aminohexanoyl)sphingosine

Synonym(s):

N-([7-Nitrobenz-2-oxa-1,3-diazol-4-yl]aminocaproyl)sphingosine, NBD hexanoic ceramide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H49N5O6
CAS Number:
Molecular Weight:
575.74
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

form

powder

lipid type

sphingolipids

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCC\C=C\[C@@H](O)[C@H](CO)NC(=O)CCCCCNc1ccc(c2nonc12)N(=O)=O

InChI

1S/C30H49N5O6/c1-2-3-4-5-6-7-8-9-10-11-12-13-15-18-27(37)25(23-36)32-28(38)19-16-14-17-22-31-24-20-21-26(35(39)40)30-29(24)33-41-34-30/h15,18,20-21,25,27,31,36-37H,2-14,16-17,19,22-23H2,1H3,(H,32,38)/b18-15+/t25-,27+/m0/s1

InChI key

HZIRBXILQRLFIK-VPZZKNKNSA-N

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tatsuru Togo
Journal of cell science, 119(Pt 13), 2780-2786 (2006-06-15)
Resealing of a disrupted plasma membrane requires Ca(2+)-regulated exocytosis. Repeated disruptions reseal more quickly than the initial wound. This facilitated response requires both Ca(2+) and protein kinase C (PKC), and is sensitive to brefeldin A. There is also evidence that
K Wolf et al.
Cellular microbiology, 3(3), 145-152 (2001-03-22)
Chlamydia pneumoniae is a bacterial obligate intracellular parasite with a developmental cycle common to all members of the genus Chlamydia. Like other chlamydiae, the developmental cycle of C. pneumoniae occurs entirely within a membrane-bound intracellular vacuole, termed an inclusion, that
Boris Boyanovsky et al.
The Journal of biological chemistry, 278(29), 26992-26999 (2003-05-02)
Ceramide is a bioactive molecule involved in cellular responses to stress and inflammation. The major pathway for ceramide accumulation is via agonist-induced activation of cellular sphingomyelinases. It has also been shown that the ceramide level in circulating low density lipoprotein
E H Pap et al.
Experimental cell research, 265(2), 288-293 (2001-04-17)
Peptides carrying organelle-specific import or retention sequences can target the fluorophore BODIPY(581/591) to the nucleus, peroxisomes, endoplasmic reticulum (ER), or the trans-Golgi network (TGN). The peroxisomal peptide contains the PTS1 sequence AKL. For targeting to the ER or TGN, the
Daniel G Milis et al.
Molecular and cellular biochemistry, 283(1-2), 57-66 (2006-01-31)
The influence of sterol carrier protein-2 (SCP-2) on the cellular metabolism of sphingolipids was examined in control mouse L-cells and stably transfected clones expressing the protein SCP-2. Three approaches were used to examine for differences; (1) compositional analysis of endogenous

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service