N8267
Nitrocellulose membranes for blotting
pore size 0.45 μm, size 15 cm × 15 cm
About This Item
Recommended Products
size
15 cm × 15 cm
pore size
0.45 μm
SMILES string
OC[C@@H]1O[C@@H](O[C@H]2[C@@H](O)[C@H](O)[C@@H](O)O[C@H]2CO)[C@@H](O)[C@H](O)[C@H]1O.[O-][N+](=O)OC[C@@H]3O[C@H](O[N+]([O-])=O)[C@@H](O[N+]([O-])=O)[C@H](O[N+]([O-])=O)[C@@H]3O[C@H]4O[C@@H](CO[N+]([O-])=O)[C@@H](O[N+]([O-])=O)[C@@H](O[N+]([O-])=O)[C@@H]4O[N+]([O-])=O
Looking for similar products? Visit Product Comparison Guide
Application
recommended
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service