Recognizes an epitope with the sequence 359-365 of the α1 subunit of the nicotinic acetylcholine receptor. Binds to both native and denatured α1 subunit.
Immunogen
denatured nicotinic acetylcholine receptors from the electric eel Torpedo.
Physical form
Solution in 10 mM phosphate buffered saline, containing 10 mM sodium azide.
Legal Information
Sold with the permission of the Salk Institute
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.