Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

N7529

Sigma-Aldrich

Anti-Cyclic Nucleotide-Gated Cation Channel 2 antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-CNG 2, Anti-OCNC1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

rat

technique(s)

western blot: suitable

UniProt accession no.

storage temp.

−20°C

Gene Information

Immunogen

synthetic peptide corresponding to amino acids 643-660 of rat CNG2 with additional N-terminal cysteine.

Physical form

Lyophilized from phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin, 5% sucrose and 0.025% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

hcodes

pcodes

Hazard Classifications

Aquatic Chronic 3

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service