N6663
Monoclonal Anti-NUP153 antibody produced in rat
~1.0 mg/mL, clone R3G1, purified immunoglobulin, buffered aqueous solution
Synonym(s):
Anti-NUCZINK, Anti-Nucleoporin 153
About This Item
biological source
rat
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
R3G1, monoclonal
form
buffered aqueous solution
mol wt
antigen apparent mol wt ~160 kDa
antigen predicted mol wt ~153 kDa
species reactivity
rat, primate, mouse, human
packaging
antibody small pack of 25 μL
concentration
~1.0 mg/mL
technique(s)
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: 2-4 μg/mL using HeLa total cell extract
isotype
IgG2a
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... NUP153(9972)
mouse ... Nup153(218210)
rat ... Nup153(25281)
General description
Immunogen
Application
Biochem/physiol Actions
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
10 - Combustible liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service